Product Details

SNP ID
rs2163005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79902138 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCTTTGTTCACTCAGGAAATAT[C/T]CACTGAGCATTGGCCACATGCCAGT
Phenotype
MIM: 604197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTHFS PubMed Links

Gene Details

Gene
MTHFS
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
There are no transcripts associated with this gene.

Gene
ST20
Gene Name
suppressor of tumorigenicity 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100879.1 Intron NP_001094349.1
NM_001100880.2 Intron NP_001094350.1
NM_001199757.1 Intron NP_001186686.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 Intron NP_001186689.1

View Full Product Details