Product Details

SNP ID
rs2176191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227610157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATTCTCAGAAACACACCTATTT[G/T]CAAAATGCAGGTGATTACATTTGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf83 PubMed Links

Gene Details

Gene
C2orf83
Gene Name
chromosome 2 open reading frame 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162483.1 2020 UTR 3 NP_001155955.1
NM_020161.3 2020 Intron NP_064546.3
Gene
LOC729968
Gene Name
spore coat protein SP96
There are no transcripts associated with this gene.

View Full Product Details