Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317231.1 | 1334 | Intron | NP_001304160.1 | ||
NM_001317232.1 | 1334 | Intron | NP_001304161.1 | ||
NM_001317233.1 | 1334 | Intron | NP_001304162.1 | ||
NM_014342.3 | 1334 | Intron | NP_055157.1 | ||
XM_006718172.2 | 1334 | Intron | XP_006718235.1 | ||
XM_011519959.2 | 1334 | UTR 3 | XP_011518261.1 | ||
XM_011519960.2 | 1334 | UTR 3 | XP_011518262.1 | ||
XM_011519961.2 | 1334 | UTR 3 | XP_011518263.1 | ||
XM_017017462.1 | 1334 | Intron | XP_016872951.1 | ||
XM_017017463.1 | 1334 | Intron | XP_016872952.1 |