Product Details

SNP ID
rs2213165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5268447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGGTGTCAGGGTCACAGGAA[C/G]ACCTGCAAACTGGAAGAGAACTCAG
Phenotype
MIM: 142100
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HBE1 PubMed Links

Gene Details

Gene
HBE1
Gene Name
hemoglobin subunit epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005330.3 719 UTR 3 NP_005321.1

View Full Product Details