Product Details

SNP ID
rs2074422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60842499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCGAAGCCAAGTGAGGAGGCACC[A/G]AAGTGTTCTCAGGACCAGGGAGTAC
Phenotype
MIM: 611293 MIM: 604837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC86 PubMed Links

Gene Details

Gene
CCDC86
Gene Name
coiled-coil domain containing 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024098.3 Intron NP_077003.1
Gene
PTGDR2
Gene Name
prostaglandin D2 receptor 2
There are no transcripts associated with this gene.

View Full Product Details