Product Details

SNP ID
rs2911708
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:217430197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCACCTTCTAAATGGGTTGGG[C/T]CATTGCCAGAAATCTGAGTTCTGGG
Phenotype
MIM: 616836
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPATCH2 PubMed Links

Gene Details

Gene
GPATCH2
Gene Name
G-patch domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297754.1 2666 Intron NP_001284683.1
NM_018040.3 2666 UTR 3 NP_060510.1
XM_011509689.2 2666 Intron XP_011507991.1
XM_011509690.2 2666 Intron XP_011507992.1
XM_011509691.2 2666 Intron XP_011507993.1
XM_011509693.2 2666 Intron XP_011507995.1
XM_011509694.2 2666 Intron XP_011507996.1
XM_017001592.1 2666 Intron XP_016857081.1
XM_017001593.1 2666 Intron XP_016857082.1

View Full Product Details