Product Details

SNP ID
rs2075291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116790676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGGAAGAGCTCTTTGAAGCGGC[A/C]GGTGTGGTGCACCACGCGGCTCTGC
Phenotype
MIM: 606368 MIM: 603901
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOA5 PubMed Links

Gene Details

Gene
APOA5
Gene Name
apolipoprotein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166598.1 626 Missense Mutation NP_001160070.1
NM_052968.4 626 Missense Mutation NP_443200.2
XM_017017169.1 626 Missense Mutation XP_016872658.1
Gene
ZPR1
Gene Name
ZPR1 zinc finger
There are no transcripts associated with this gene.

View Full Product Details