Product Details
- SNP ID
-
rs2228019
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:70598344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAATCCAGCCGCACTGACCAGAT[A/G]GGGTTGTTATTGTCCCACACGGTGC
- Phenotype
-
MIM: 170280
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRF1
PubMed Links
Gene Details
- Gene
- PRF1
- Gene Name
- perforin 1
View Full Product Details