Product Details

SNP ID
rs2229135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5953089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGACTTAGCTGGGCTGGTTTCCC[C/T]GAGTTTCATAGGTGGTGAGAGCAGT
Phenotype
MIM: 601070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL15RA PubMed Links

Gene Details

Gene
IL15RA
Gene Name
interleukin 15 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243539.1 1323 UTR 3 NP_001230468.1
NM_001256765.1 1323 UTR 3 NP_001243694.1
NM_002189.3 1323 UTR 3 NP_002180.1
NM_172200.2 1323 UTR 3 NP_751950.2
XM_011519461.2 1323 Intron XP_011517763.1
XM_011519462.2 1323 Intron XP_011517764.1
XM_011519463.2 1323 Intron XP_011517765.1
XM_011519464.2 1323 Intron XP_011517766.1
XM_011519465.1 1323 UTR 3 XP_011517767.1
XM_011519466.2 1323 UTR 3 XP_011517768.1
XM_011519467.2 1323 Intron XP_011517769.1
XM_011519468.1 1323 Intron XP_011517770.1
XM_011519469.1 1323 UTR 3 XP_011517771.1
XM_011519470.2 1323 Intron XP_011517772.1
XM_011519471.2 1323 Intron XP_011517773.1
XM_011519472.2 1323 Intron XP_011517774.1
XM_011519474.2 1323 Intron XP_011517776.1
XM_011519475.2 1323 Intron XP_011517777.1
XM_011519476.2 1323 UTR 3 XP_011517778.1
XM_011519477.2 1323 UTR 3 XP_011517779.1
XM_017016195.1 1323 UTR 3 XP_016871684.1
XM_017016196.1 1323 Intron XP_016871685.1
XM_017016197.1 1323 Intron XP_016871686.1
XM_017016198.1 1323 UTR 3 XP_016871687.1
XM_017016199.1 1323 Intron XP_016871688.1
XM_017016200.1 1323 Intron XP_016871689.1
XM_017016201.1 1323 UTR 3 XP_016871690.1
XM_017016202.1 1323 UTR 3 XP_016871691.1
XM_017016203.1 1323 UTR 3 XP_016871692.1

View Full Product Details