Product Details

SNP ID
rs2243128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:159992724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCAAGTGTCACAGTGAGGTGT[A/G]GAGGCTGGTTAGCACAGGCTGTTGA
Phenotype
MIM: 161560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL12A PubMed Links

Gene Details

Gene
IL12A
Gene Name
interleukin 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000882.3 Intron NP_000873.2
Gene
IL12A-AS1
Gene Name
IL12A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details