Product Details

SNP ID
rs2286796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15980913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAGAGGTGACTAACTGAAGACAA[A/G]GACAAAAAACTACCATTTGTTTCAA
Phenotype
MIM: 600446 MIM: 614535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
There are no transcripts associated with this gene.

Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 Intron NP_001036162.1
NM_001042698.1 Intron NP_001036163.1

View Full Product Details