Product Details

SNP ID
rs2287578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171605515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCTCTATATATGCAAATATATA[C/T]ATGCAAATTATTTGAGAGACCATGA
Phenotype
MIM: 602382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLD1 PubMed Links

Gene Details

Gene
PLD1
Gene Name
phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130081.2 Intron NP_001123553.1
NM_002662.4 Intron NP_002653.1
XM_005247533.2 Intron XP_005247590.1
XM_005247534.2 Intron XP_005247591.1
XM_011512897.1 Intron XP_011511199.1
XM_011512898.1 Intron XP_011511200.1
XM_011512899.2 Intron XP_011511201.1
XM_017006623.1 Intron XP_016862112.1
XM_017006624.1 Intron XP_016862113.1

View Full Product Details