Product Details
- SNP ID
-
rs2297298
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:60092494 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCCAAGCCTGTGCGGAGAGCCAAG[A/T]TGTTCTAGGGGCTTTCATTGCCACA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PCNX4
PubMed Links
Gene Details
- Gene
- PCNX4
- Gene Name
- pecanex homolog 4 (Drosophila)
View Full Product Details