Product Details
- SNP ID
-
rs2298063
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
39
- Location
-
Chr.1:223394189 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- CCTCCAGCAAAGAGATGCGGAGCCC[A/G]CACACCCAGGTCCTGAAGAGCAAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC185
PubMed Links
Gene Details
- Gene
- CCDC185
- Gene Name
- coiled-coil domain containing 185
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152610.2 |
817 |
Silent Mutation |
CCA,CCG |
P238P |
NP_689823.2 |
View Full Product Details