Product Details

SNP ID
rs2294985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62818037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGGGCTTAGGAGCCGACTCA[A/G]TCCTGAGATGATGTCCCCTATGGGT
Phenotype
MIM: 120270 MIM: 606459
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL9A3 PubMed Links

Gene Details

Gene
COL9A3
Gene Name
collagen type IX alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001853.3 Intron NP_001844.3
XM_011528545.1 Intron XP_011526847.1
XM_017027666.1 Intron XP_016883155.1
Gene
OGFR
Gene Name
opioid growth factor receptor
There are no transcripts associated with this gene.

View Full Product Details