Product Details

SNP ID
rs2298383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24429543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGATCCTGGCCCCGCCCCCTCCC[C/T]GGGGCCTCTGACCTTCCTGTCACTC
Phenotype
MIM: 102776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA2A PubMed Links

Gene Details

Gene
ADORA2A
Gene Name
adenosine A2a receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000675.5 Intron NP_000666.2
NM_001278497.1 Intron NP_001265426.1
NM_001278498.1 Intron NP_001265427.1
NM_001278499.1 Intron NP_001265428.1
NM_001278500.1 Intron NP_001265429.1
Gene
ADORA2A-AS1
Gene Name
ADORA2A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SPECC1L-ADORA2A
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details