Product Details

SNP ID
rs2298437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30496968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCCATAGCATGATGGGCGGCAG[C/T]AGCCATATCCGTTGCCTCCAAAGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-2 PubMed Links

Gene Details

Gene
KRTAP19-2
Gene Name
keratin associated protein 19-2
There are no transcripts associated with this gene.

Gene
KRTAP19-3
Gene Name
keratin associated protein 19-3
There are no transcripts associated with this gene.

Gene
KRTAP19-4
Gene Name
keratin associated protein 19-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181610.1 143 Missense Mutation TAC,TGC Y48C NP_853641.1
Gene
KRTAP19-5
Gene Name
keratin associated protein 19-5
There are no transcripts associated with this gene.

View Full Product Details