Product Details
- SNP ID
-
rs2295598
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116571676 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CACAGGTGACTCCCGCGGCCGCAAG[C/T]CTGTAGCCGCCCCGAGGTGGAGTAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD1
PubMed Links
Gene Details
- Gene
- RWDD1
- Gene Name
- RWD domain containing 1
View Full Product Details