Product Details

SNP ID
rs2295777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100535889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGTTGGGTGTGTGGTTTTTTTG[G/T]TTTTTTTTTTTTGGGGTCCGGAATA
Phenotype
MIM: 606615 MIM: 602140
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIF1AN PubMed Links

Gene Details

Gene
HIF1AN
Gene Name
hypoxia inducible factor 1 alpha subunit inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017902.2 6 UTR 5 NP_060372.2
XM_011539940.2 6 Intron XP_011538242.1
XM_011539941.1 6 Intron XP_011538243.1
Gene
NDUFB8
Gene Name
NADH:ubiquinone oxidoreductase subunit B8
There are no transcripts associated with this gene.

View Full Product Details