Product Details

SNP ID
rs2295799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35821800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACTGGCAACTATACTTCAGATT[C/G]TGTTACAAGTGTTAGTGCTCTTAAA
Phenotype
MIM: 609997 MIM: 605731 MIM: 616888
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM221B PubMed Links

Gene Details

Gene
FAM221B
Gene Name
family with sequence similarity 221 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012446.3 Intron NP_001012448.2
XM_006716768.3 Intron XP_006716831.1
Gene
HINT2
Gene Name
histidine triad nucleotide binding protein 2
There are no transcripts associated with this gene.

Gene
SPAG8
Gene Name
sperm associated antigen 8
There are no transcripts associated with this gene.

Gene
TMEM8B
Gene Name
transmembrane protein 8B
There are no transcripts associated with this gene.

View Full Product Details