Product Details

SNP ID
rs2306445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70198247 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGAATCAGCTTTGAAATAGGCAG[A/G]GGTTTTTGAACAATATTTTTTCTCA
Phenotype
MIM: 614843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ODAM PubMed Links

Gene Details

Gene
ODAM
Gene Name
odontogenic, ameloblast asssociated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017855.3 Intron NP_060325.3
XM_005265698.3 Intron XP_005265755.1
XM_006714245.3 Intron XP_006714308.1
XM_017008331.1 Intron XP_016863820.1

View Full Product Details