Product Details

SNP ID
rs2307160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42338563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAAGTCCTGGTACCTCCTTCA[A/G]GCTGGGAGAGGGCTCTAGTCCCTGG
Phenotype
MIM: 603258 MIM: 174760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKBKB PubMed Links

Gene Details

Gene
IKBKB
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
There are no transcripts associated with this gene.

Gene
POLB
Gene Name
DNA polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002690.2 106 UTR 5 NP_002681.1
XM_005273535.3 106 UTR 5 XP_005273592.1
XM_005273536.3 106 UTR 5 XP_005273593.1
XM_005273537.3 106 UTR 5 XP_005273594.1
XM_005273538.2 106 UTR 5 XP_005273595.1
XM_005273539.2 106 UTR 5 XP_005273596.1
XM_005273540.4 106 UTR 5 XP_005273597.1
XM_006716353.2 106 Intron XP_006716416.1
XM_017013583.1 106 UTR 5 XP_016869072.1
XM_017013584.1 106 UTR 5 XP_016869073.1

View Full Product Details