Product Details

SNP ID
rs2307301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24777319 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGTATGTGATTGAATAACTTTA[A/T]TTTTTTTTTGTAGAAAGCATGGGGC
Phenotype
MIM: 602842
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GMNN PubMed Links

Gene Details

Gene
GMNN
Gene Name
geminin, DNA replication inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251989.1 Intron NP_001238918.1
NM_001251990.1 Intron NP_001238919.1
NM_001251991.1 Intron NP_001238920.1
NM_015895.4 Intron NP_056979.1
XM_005249159.1 Intron XP_005249216.1
XM_011514651.2 Intron XP_011512953.1

View Full Product Details