Product Details
- SNP ID
-
rs2327264
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:10225606 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTTGCTGTTGTTGATATGGTTCA[C/T]ATTACTCAAAGATGTCTAATTCTCA
- Phenotype
-
MIM: 600322
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SNAP25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150267082] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNAP25
- Gene Name
- synaptosome associated protein 25
- Gene
- SNAP25-AS1
- Gene Name
- SNAP25 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details