Product Details
- SNP ID
-
rs3026286
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:90063655 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTGTCCATTGCTAGAGTTTATCAG[A/G]ATAATATAGAATAGTCAAACAGAGA
- Phenotype
-
MIM: 114050
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALB1
PubMed Links
Gene Details
- Gene
- CALB1
- Gene Name
- calbindin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004929.3 |
|
Intron |
|
|
NP_004920.1 |
View Full Product Details