Product Details
- SNP ID
-
rs2244053
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:5892048 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AACTTTGGAGAAAAATAACTCCTCT[A/T]ATTTTGTTAGCCACAGCCCTGGGCC
- Phenotype
-
MIM: 605513
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2026
PubMed Links
Gene Details
- Gene
- KIAA2026
- Gene Name
- KIAA2026
- Gene
- MLANA
- Gene Name
- melan-A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005511.1 |
7230 |
Intron |
|
|
NP_005502.1 |
View Full Product Details