Product Details

SNP ID
rs2466759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47776935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCAATTTTTTTCAGCAACATTTA[C/T]TTCTTGAATCCATGACCCTCCTTTT
Phenotype
MIM: 600899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKDC PubMed Links

Gene Details

Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081640.1 12055 Missense Mutation ATA,GTA I4000V NP_001075109.1
NM_006904.6 12055 Missense Mutation ATA,GTA I4031V NP_008835.5

View Full Product Details