Product Details
- SNP ID
-
rs2460052
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:94024229 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGGAAAGAACCTGAAATAGAAAT[C/T]CAGAGACCTAACTTCTAATGCTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HEPHL1
PubMed Links
Gene Details
- Gene
- HEPHL1
- Gene Name
- hephaestin like 1
View Full Product Details