Product Details

SNP ID
rs2531217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2040403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACATGTGACCATCTTGGCTGCCC[A/C]TGAGGTGCTGGGGCAGGGGCTGAGC
Phenotype
MIM: 602656 MIM: 606553 MIM: 191092
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NTHL1 PubMed Links

Gene Details

Gene
NTHL1
Gene Name
nth-like DNA glycosylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318193.1 Intron NP_001305122.1
NM_001318194.1 Intron NP_001305123.1
NM_002528.6 Intron NP_002519.1
XM_017023253.1 Intron XP_016878742.1
Gene
SLC9A3R2
Gene Name
SLC9A3 regulator 2
There are no transcripts associated with this gene.

Gene
TSC2
Gene Name
tuberous sclerosis 2
There are no transcripts associated with this gene.

View Full Product Details