Product Details

SNP ID
rs2576052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47163736 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATGAGCTTTTTGACTTTGAAAA[C/T]CATACACATATTACTTTGATAAAAG
Phenotype
MIM: 609382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IER3IP1 PubMed Links

Gene Details

Gene
IER3IP1
Gene Name
immediate early response 3 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016097.4 Intron NP_057181.1

View Full Product Details