Product Details

SNP ID
rs2664369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:88037303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGGTACACAGTATATGAAATG[G/T]CCAAATTATGTGTTAACAAAAAAAG
Phenotype
MIM: 602262
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MMP16 PubMed Links

Gene Details

Gene
MMP16
Gene Name
matrix metallopeptidase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005941.4 6264 UTR 3 NP_005932.2

View Full Product Details