Product Details

SNP ID
rs2706235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50717859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCAAGCACCTAGAATGACATTT[A/G]CAAAGCAGGCAAAATGAAAATGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLEU7 PubMed Links

Gene Details

Gene
DLEU7
Gene Name
deleted in lymphocytic leukemia, 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306135.1 Intron NP_001293064.1
NM_198989.3 Intron NP_945340.2

View Full Product Details