Product Details

SNP ID
rs2741514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45294011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCTCTGTCCCTGCGCTGATGCC[C/T]TCCTCTGCAAGCCGGACACAGAGGG
Phenotype
MIM: 603897
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MATN4 PubMed Links

Gene Details

Gene
MATN4
Gene Name
matrilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003833.4 1764 Silent Mutation GAA,GAG E528E NP_003824.2
NM_030590.3 1764 Silent Mutation GAA,GAG E487E NP_085080.1
NM_030592.3 1764 Silent Mutation GAA,GAG E446E NP_085095.1
XM_005260597.1 1764 Silent Mutation GAA,GAG E528E XP_005260654.1
XM_017028113.1 1764 Silent Mutation GAA,GAG E528E XP_016883602.1
XM_017028114.1 1764 Silent Mutation GAA,GAG E487E XP_016883603.1
XM_017028115.1 1764 Silent Mutation GAA,GAG E446E XP_016883604.1

View Full Product Details