Product Details

SNP ID
rs2725234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88011120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACCAAAGGAAGCCAAGGATGAC[C/T]GAAAGATTTAAAGATGAAGCCCAGA
Phenotype
MIM: 173910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKD2 PubMed Links

Gene Details

Gene
PKD2
Gene Name
polycystin 2, transient receptor potential cation channel
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000297.3 Intron NP_000288.1
XM_011532028.2 Intron XP_011530330.1
XM_011532029.1 Intron XP_011530331.1
XM_011532030.2 Intron XP_011530332.1

View Full Product Details