Product Details
- SNP ID
-
rs2737194
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:117704453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGGGCCACTGCTGCTCACAGAA[C/G]CAGTGAGGATGATGCCAGGATGATG
- Phenotype
-
MIM: 603030
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TLR4
PubMed Links
Gene Details
- Gene
- TLR4
- Gene Name
- toll like receptor 4
View Full Product Details