Product Details

SNP ID
rs2758910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39299539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGGGGAGTGGGACTCTGGCTCC[C/T]GGTCAGGTCCCTGTCTCCAGCTTTG
Phenotype
MIM: 607370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK17 PubMed Links

Gene Details

Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135111.1 1179 UTR 3 NP_001128583.1
NM_031460.3 1179 Missense Mutation CAG,CGG Q296R NP_113648.2

View Full Product Details