Product Details

SNP ID
hCV22271872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24780791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTGGTGATACAGTGTCTACATC[C/G]AAAACATTTCTACTATTTTCTTGGT
Phenotype
MIM: 602842
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GMNN PubMed Links

Gene Details

Gene
GMNN
Gene Name
geminin, DNA replication inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251989.1 Intron NP_001238918.1
NM_001251990.1 Intron NP_001238919.1
NM_001251991.1 Intron NP_001238920.1
NM_015895.4 Intron NP_056979.1

View Full Product Details