Product Details

SNP ID
hCV22272961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143513850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGTTTATTGCGACGTAGCAT[C/T]CAGGCCTAGGCCTCCCCGTCCATCC
Phenotype
MIM: 603375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMARCA5 PubMed Links

Gene Details

Gene
SMARCA5
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003601.3 388 UTR 5 NP_003592.3
Gene
SMARCA5-AS1
Gene Name
SMARCA5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details