Product Details
- SNP ID
-
rs34683072
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53289701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTCTTTGCTTGAAATCAATAACC[A/G]CGCTGCACATGGGACATCTGTCAAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35700345] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BIRC8
- Gene Name
- baculoviral IAP repeat containing 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033341.4 |
|
Intron |
|
|
NP_203127.3 |
- Gene
- FAM90A27P
- Gene Name
- family with sequence similarity 90, member A27, pseudogene
There are no transcripts associated with this gene.
View Full Product Details