Product Details
- SNP ID
-
rs4880083
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137050284 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCACCCGCCTGCCCCTACTCAC[C/G]ACAAAGTTCCCAGCCACTGGGGGCT
- Phenotype
-
MIM: 602012
MIM: 605798
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ENTPD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10870170,rs113918890] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ENTPD2
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 2
- Gene
- NPDC1
- Gene Name
- neural proliferation, differentiation and control 1
There are no transcripts associated with this gene.
View Full Product Details