Product Details

SNP ID
rs4759058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53945268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACTCCACCTGACCACCCACCCG[A/C]TGCTTGCCCCATCTATTTATGTCTC
Phenotype
MIM: 142975 MIM: 142976
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXC12 PubMed Links

Gene Details

Gene
HOXC12
Gene Name
homeobox C12
There are no transcripts associated with this gene.

Gene
HOXC13
Gene Name
homeobox C13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017410.2 1120 UTR 3 NP_059106.2
Gene
HOXC13-AS
Gene Name
HOXC13 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details