Product Details

SNP ID
rs3827426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72275494 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGATCAAGACAGTATTTGGATA[C/T]CATAGGGCTGCCATCAATATGCGTC
Phenotype
MIM: 312760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS4X PubMed Links

Gene Details

Gene
RPS4X
Gene Name
ribosomal protein S4, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007.4 Intron NP_000998.1

View Full Product Details