Product Details

SNP ID
rs17597773
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:220881419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCCTAACGGGCGAGCCGACTAA[C/G]AGTCAGAAATCTCAGGACTGACGCC
Phenotype
MIM: 142995
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLX PubMed Links

Gene Details

Gene
HLX
Gene Name
H2.0 like homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021958.3 Intron NP_068777.1
Gene
HLX-AS1
Gene Name
HLX antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details