Product Details

SNP ID
rs73590153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49365397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCAGAGGGCAGAAAGTAAAGGG[A/G]ATGGGAGGGACTTGGGGCAAGGCCT
Phenotype
MIM: 605418 MIM: 601729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKKL1 PubMed Links

Gene Details

Gene
DKKL1
Gene Name
dickkopf like acrosomal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197301.1 Intron NP_001184230.1
NM_001197302.1 Intron NP_001184231.1
NM_014419.3 Intron NP_055234.1
XM_006723142.2 Intron XP_006723205.1
XM_011526724.1 Intron XP_011525026.1
XM_011526725.1 Intron XP_011525027.1
XM_011526726.2 Intron XP_011525028.1
XM_011526727.2 Intron XP_011525029.1
XM_011526728.2 Intron XP_011525030.1
XM_011526729.1 Intron XP_011525031.1
XM_017026593.1 Intron XP_016882082.1
Gene
LOC101928295
Gene Name
uncharacterized LOC101928295
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details