Product Details

SNP ID
rs9919624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112088838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAAGATGTGTGTTTCTCACATC[A/G]ACTTTTATGAATCTGGTCCTTTTTG
Phenotype
MIM: 602690 MIM: 606659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf57 PubMed Links

Gene Details

Gene
C11orf57
Gene Name
chromosome 11 open reading frame 57
There are no transcripts associated with this gene.

Gene
SDHD
Gene Name
succinate dehydrogenase complex subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276503.1 Intron NP_001263432.1
NM_001276504.1 Intron NP_001263433.1
NM_001276506.1 Intron NP_001263435.1
NM_003002.3 Intron NP_002993.1
Gene
TIMM8B
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
There are no transcripts associated with this gene.

View Full Product Details