Product Details

SNP ID
rs62191400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181457912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCGGCGTGAGAATGGCGCCCT[A/G]GGGATTCCCTGCCCGATTCAAACTT
Phenotype
MIM: 192975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA4 PubMed Links

Gene Details

Gene
ITGA4
Gene Name
integrin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000885.5 Intron NP_000876.3
NM_001316312.1 Intron NP_001303241.1

View Full Product Details