Product Details

SNP ID
rs77259832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95022464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAAACCAGAAAGAAATTTAGTA[G/T]AAAAATATGAGTAATTTCTGCATGT
Phenotype
MIM: 601529
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NR2C1 PubMed Links

Gene Details

Gene
NR2C1
Gene Name
nuclear receptor subfamily 2 group C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032287.2 Intron NP_001027458.1
NM_001127362.1 Intron NP_001120834.1
NM_003297.3 Intron NP_003288.2
XM_005269133.2 Intron XP_005269190.1
XM_006719584.2 Intron XP_006719647.1
XM_006719585.2 Intron XP_006719648.1
XM_011538716.2 Intron XP_011537018.1
XM_011538717.2 Intron XP_011537019.1

View Full Product Details