Product Details

SNP ID
rs34879346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43461534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTCCTCATCCCGGGAGGCCTC[C/G]TGTTCTACTCCCTGTCTCGGAGTCT
Phenotype
MIM: 609484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYPD3 PubMed Links

Gene Details

Gene
LYPD3
Gene Name
LY6/PLAUR domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014400.2 966 Silent Mutation CAC,CAG H286Q NP_055215.2

View Full Product Details