Product Details
- SNP ID
-
hCV25594378
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41724998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTTCCATCACCCCTTCCTCAGC[C/T]GGCACCTTCTTGGCAGCCCCCAGCT
- Phenotype
-
MIM: 137250
MIM: 600947
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAST
PubMed Links
Gene Details
- Gene
- GAST
- Gene Name
- gastrin
There are no transcripts associated with this gene.
- Gene
- HAP1
- Gene Name
- huntingtin associated protein 1
There are no transcripts associated with this gene.
View Full Product Details