Product Details
- SNP ID
-
hCV25594633
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:34709928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGGGATTCACAGGGAGCCAGGG[A/G]CTGCTGGCAAGCTCCTATCCAGGCC
- Phenotype
-
MIM: 602737
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCL21
PubMed Links
Gene Details
- Gene
- CCL21
- Gene Name
- C-C motif chemokine ligand 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002989.3 |
|
Intron |
|
|
NP_002980.1 |
View Full Product Details